Bestelling invoeren
Netherlands
ContactUsLinkComponent
(±)-Camphorquinone for synthesis, Sigma-Aldrich®
(±)-Camphorquinone for synthesis, Sigma-Aldrich®
Catalog # 8.14531.0010
Supplier:  Merck
CAS Number:  
(±)-Camphorquinone for synthesis, Sigma-Aldrich®
Catalog # 8.14531.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    198 - 201 °C
  • Molar Mass:
    166.22 g/mol
  • Synonyms:
    2,3-Bornanedione
  • Hill Formula:
    C10H14O2

Specification Test Results

About this item

 :
 :166.22 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:233-814-1
CAS Number:10373-78-1
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00064160