Order Entry
Netherlands
ContactUsLinkComponent
 
Flammable
Irritant
Dimethoxy(diisopropyl)silane for synthesis, Sigma-Aldrich®
Catalog # 8.14719.0025
Supplier:  Merck
CAS Number:  
Flammable
Irritant
Dimethoxy(diisopropyl)silane for synthesis, Sigma-Aldrich®
Catalog # 8.14719.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    176.33 g/mol
  • Boiling Point:
    164 °C (1013 hPa)
  • Vapor Pressure:
    2.14 hPa (20 °C)
  • Flash Point:
    44 °C
  • Density:
    0.88 g/cm3 (20 °C)
  • Ignition Point:
    240 °C

Specification Test Results

About this item

 :
 :176.33 g/mol
ADR:3,III
 :M-Clarity™ quality level = MQ200
EINECS:421-540-7
UN:1993
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:18230-61-0
Formula:C₈H₂₀O₂Si
MDL Number:MFCD00239343