Supelco® Organic Reference Standard, Trichloroethylene
46267-5ML-R
: Merck
:
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:79-01-6
- Description:Trichloroethylene
- Seq:2210
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00000838
- M-Clarity quality level:MQ100
- Synonym(s):TCE; Trichloroethene
- Melting point:–84.8 °C (lit.)
- Formula:ClCH=CCl₂
- Molecular weight:131,39
- Density:1,463 g/ml at 25 °C (lit.)
- Flash point:Does not flash
- Boiling point:86.7 °C (lit.)
- GHS signal: GHS08, GHS07