Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

2,5-Dibromothiophene 95%

2,5-Dibromothiophene 95%
 
2,5-Dibromothiophene 95%
2,5-Dibromothiophene 95%

 

 
  •  :3141-27-3
  •  :C₄H₂Br₂S
  •  :MFCD00005420
  •  :241.93 g/mol
  •  :211 °C (760 mmHg)
  •  :–6 °C
  •  :2.14 g/cm³
  •  :99 °C
  •  :Ambient
  •  :221-547-3
  •  :6.1,II

 

 

 

  • Appearance:
    Clear light yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6270 to 1.6300 (20°C, 589 nm)
  • GC:
    ≥94 %
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    25 g
       
       
    1 * 25 g
    1 * 25 g - 43,00 €
    43,00 €
    1 * 25 g
       43,00 €

  • Glass bottle for solids
    100 g
       
       
    1 * 100 g
    1 * 100 g - 127,00 €
    127,00 €
    1 * 100 g
       127,00 €

  • Glass bottle
    500 g
       
       
    1 * 500 g
    1 * 500 g - 556,00 €
    556,00 €
    1 * 500 g
       556,00 €