2-(3-Phenylpropyl)pyridine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 2110-18-1 Molecular Formula: C14H15N Formula Weight: 197.28 Flash Point: 100°(212°F) Boiling Point: 142-143°/1mm MDL No.: MFCD00191237
- CAS Number:2110-18-1
- Formula:C₁₄H₁₅N
- MDL Number:MFCD00191237
- Molecular Weight:197.28 g/mol
- Boiling Pt:142…143 °C (1 mmHg)
- Density:1.017
- Flash Pt:100 °C (212 °F)
- Storage Temperature:Ambient
- EINECS:218-300-7
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 2110-18-1 Molecular Formula: C14H15N Formula Weight: 197.28 Flash Point: 100°(212°F) Boiling Point: 142-143°/1mm MDL No.: MFCD00191237
- CAS Number:2110-18-1
- Formula:C₁₄H₁₅N
- MDL Number:MFCD00191237
- Molecular Weight:197.28 g/mol
- Boiling Pt:142…143 °C (1 mmHg)
- Density:1.017
- Flash Pt:100 °C (212 °F)
- Storage Temperature:Ambient
- EINECS:218-300-7



