
2,3-Diaminopyridine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:452-58-4
:
Formula:C₅H₇N₃
MDL Number:MFCD00006319
:109.13 g/mol
Boiling Pt:148…150 °C (5 mmHg)
Melting Pt:110…112 °C
Storage Temperature:Ambient
EINECS:207-200-9
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS07
Specification Test Results
- Melting Point:108 - 113 °C
- Molar Mass:109.13 g/mol
Specification Test Results
Frequently Bought Together