Order Entry
Netherlands
ContactUsLinkComponent
Tetrahydrofuran
Flammable
Health
Irritant
Tetrahydrofuran
Catalog # ROTHAE07.1
Supplier:  Roth Carl
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran
Catalog # ROTHAE07.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356