


Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–21.5 °C
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
Related Information
Certificates
Search by Lot NumberMERC1.08114.1000_TDS_EN MERC1.08114.6190_TDS_EN ProductPage_108114 ProductPage_108114
Product specification
msds_108114
SDS
Frequently Bought Together