


Naphthalene
Catalog # PROOERM-FC028
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:128.17 g/mol
Merck Index:12,06457
Boiling Pt:218 °C (1013 hPa)
Melting Pt:79…82 °C
Density:1.145 g/cm³ (20 °C)
Flash Pt:79 °C
EINECS:202-049-5
UN:1334
ADR:4.1,III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:91-20-3
Formula:C₁₀H₈
MDL Number:MFCD00001742