
1,4-Dimethoxynaphthalene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:10075-62-4
:
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00052378
:188.23 g/mol
EINECS:233-209-2
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together