Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octachloronaphthalene
Octachloronaphthalene
Octachloronaphthalene
Octachloronaphthalene
PROOCIL-ECN-5280
:
LGC Standards PROMOCHEM
:
Octachloronaphthalene
PROOCIL-ECN-5280
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
Seq:
2080
Environmentally Preferable:
:
:
403.73 g/mol
:
218-778-7
:
GHS07
:
2234-13-1
:
C₁₀Cl₈