Order Entry
Netherlands
ContactUsLinkComponent
1-Chloronaphthalene 85% remainder 2-Chloronaphthalene, Technical Grade
Irritant
Environment
1-Chloronaphthalene 85% remainder 2-Chloronaphthalene, Technical Grade
Certificates
undefined
Irritant
Environment
1-Chloronaphthalene 85% remainder 2-Chloronaphthalene, Technical Grade
Certificates

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6300 to 1.6340 (20°C, 589 nm)
  • HPLC:
    ≥80 %

Specification Test Results

Related Information

Frequently Bought Together