Osmium tetraoxide
: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
:20816-12-0
:
:O₄Os
:MFCD00011150
:254.23 g/mol
:13,06962
:130 °C (1013 hPa)
:40 °C
:1.04 g/cm³ (20 °C)
:Ambient
:244-058-7
:2471
:6.1,I
:GHS06
:GHS05
- Environmentally Preferable:
Frequently Bought Together