

Zirconium tetrachloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10026-11-6
:
Formula:Cl₄Zr
MDL Number:MFCD00011306
:233.03 g/mol
Merck Index:12,10307
Boiling Pt:3577 °C (1013 hPa)
Melting Pt:437 °C
Density:2.8 g/cm³ (20 °C)
UN:2503
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:233-058-2
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:437 °C
- Molar Mass:233.04 g/mol
- Bulk Density:1200 kg/m3
- Vapor Pressure:1.3 hPa (190 °C)
- Density:2.80 g/cm3 (15 °C)
- pH:- 1 (H2O, 20 °C) (saturated aqueous solution)
Specification Test Results
Frequently Bought Together