Lanthanum(III) oxide 99.99% (trace metals basis)
:1312-81-8
:
:La₂O₃
:MFCD00011071
:325.81 g/mol
:4200 °C (760 mmHg)
:2315 °C
:6,51 g/cm³ (20 °C)
:Ambient
:215-200-5
- Appearance:White Powder
- Assay:≥99.94 % (La2O3) TREO base
- Loss on ignition:≤2.0 % (1000°C)
- Iron(III)oxide (Fe2O3):≤10 ppm
Frequently Bought Together