Orthoboric acid, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
Specification Test Results
- Assay:min. 99.5 %
- Arsenic (As):max. 1 ppm
- Calcium (Ca):max. 0.005 %
- Chloride (Cl):max. 0.001 %
- Iron (Fe):max. 0.001 %
- Heavy Metals (as Pb):max. 0.001 %
- Insoluble in Methanol:max. 0.005 %
- Nonvolatile with Methanol:max. 0.05 %
- Phosphate (PO4):max. 0.001 %
- Sulfate (SO4):max. 0.005 %
Specification Test Results
Frequently Bought Together