
Octadecyl methacrylate ≥97.0% (by GC) stabilised
: TCI
:32360-05-7
:
:C₂₂H₄₂O₂
:MFCD00026683
:338.57 g/mol
:235°C
:191°C
:Stabilised with 4-methoxyphenol (MEHQ)
:251-013-5
:GHS07
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together