3-Hydroxypicolinic acid ≥99.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
Leverancier: Merck
Certificaten
Over dit artikel
CAS-nummer:874-24-8
:
Formule:C₆H₅NO₃
MDL-nummer:MFCD00006294
:139.11 g/mol
:M-Clarity™ quality level = MQ100
EINECS:212-859-0
Frequently Bought Together
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM®, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Leverancier: VWR Chemicals
Hypodermic needles, HENKE-JECT® for single use
Leverancier: HENKE SASS WOLF
2-Propanol ≥99.0%, GPR RECTAPUR®
Leverancier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Leverancier: Merck
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Leverancier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
Leverancier: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverancier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals