Dimethyl-3-oxoglutarate
Catalog # MOLEM84420468
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:174.15 g/mol
EINECS:217-385-8
CAS Number:1830-54-2
Formula:C₇H₁₀O₅
MDL Number:MFCD00008462