



Acryloyl chloride stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:814-68-6
:
Formula:C₃H₃ClO
MDL Number:MFCD00000731
:90.51 g/mol
ADR:6.1 (3, 8),I
:M-Clarity™ quality level = MQ200
:Stabilised with phenothiazine
EINECS:212-399-0
UN:3488
:GHS02
:GHS06
:GHS05
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Molar Mass:90.5 g/mol
- Boiling Point:74 - 76 °C (1013 hPa)
- Vapor Pressure:106.6 hPa (20 °C)
- Flash Point:-4 °C
- Refractive Index:1.435 (20 °C, 589 nm)
- Density:1.12 g/cm3 (20 °C)
- pH:(H2O) acidic
Specification Test Results
Frequently Bought Together