Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Inloggen
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
Netherlands
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
ContactUsLinkComponent
Home
Acetamidine hydrochloride
Acetamidine hydrochloride
Acetamidine hydrochloride
Acetamidine hydrochloride
Catalog # 8.00026.9020
Supplier:
Merck
CAS Number:
Expand image
Acetamidine hydrochloride
Catalog # 8.00026.9020
Supplier:
Merck
CAS Number:
Specifications
Pk:
20 kg
Seq:
4241
Specifications
About this item
Product Details
:
:
94.54 g/mol
EINECS:
204-700-9
CAS Number:
124-42-5
Formula:
C₂H₇ClN₂
MDL Number:
MFCD00013016
Product Details