
Lithium sulphate, anhydrous ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10377-48-7
:
Formula:Li₂O₄S
MDL Number:MFCD00011086
:109.95 g/mol
Merck Index:14,5541
Boiling Pt:1377 °C (1013 hPa)
Melting Pt:845 °C
Density:2.22
EINECS:233-820-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together