
Sodium hydroxide ≥97%, pearls
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,8627
Boiling Pt:1390 °C
Melting Pt:318 °C
Density:2.13
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
Related Information
Frequently Bought Together