Order Entry
Netherlands
ContactUsLinkComponent
Potassium cyanate for synthesis, Sigma-Aldrich®
Irritant
Potassium cyanate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Potassium cyanate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    750 g/l (20 °C)
  • Melting Point:
    314 - 316 °C
  • Molar Mass:
    81.12 g/mol
  • Bulk Density:
    870 kg/m3
  • Density:
    2.056 g/cm3 (20 °C)
  • pH:
    10 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together