Water, HyClone™ HyPure for injection (WFI), sterile filtered
Supplier: HyClone products (Cytiva)
Certificates
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Molecular Weight:18.02 g/mol
Merck Index:13,10098
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Sterilisation:0,1 µm sterile filtered
Specification Test Results
Related Information
Frequently Bought Together