


Sodium nitrite, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7632-00-0
:
Formula:NaNO₂
MDL Number:MFCD00011118
:69 g/mol
Merck Index:13,08721
Boiling Pt:320 °C (1013 hPa)
Melting Pt:284 °C
Density:2.168 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1500
ADR:5.1 (6.1),III
:M-Clarity™ quality level = MQ300
EINECS:231-555-9
:GHS03
:GHS06
:GHS09
Specification Test Results
- Solubility:820 g/l (20 °C)
- Melting Point:280 °C (decomposition)
- Molar Mass:69.00 g/mol
- Bulk Density:1200 kg/m3
- Boiling Point:320 °C (decomposition)
- Density:2.1 g/cm3 (20 °C)
- pH:9 (100 g/l, H2O, 20 °C)
- Ignition Point:510 °C
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06549.0100_TDS_EN ProductPage_106549
Product specification
msds_106549
SDS
Frequently Bought Together