


Sodium nitrite, BAKER, Laboratory Reagent, J.T.Baker®
0298.1000
: Avantor
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Assay (dried basis):98.5 - 100.5 %
- Acidity or Alkalinity:passes test
- Appearance of solution:passes test
- Chloride (Cl):max. 50 ppm
- Heavy Metals (as Pb):max. 20 ppm
- Identification:passes test
- Loss on Drying:max. 0.25 %
- Sulfate (SO4):max. 200 ppm
:
:69 g/mol
:13,08721
:320 °C (1013 hPa)
:284 °C
:2.168 g/cm³ (20 °C)
:Ambient
:231-555-9
:1500
:5.1,III
:GHS03
:GHS06
:GHS09
:7632-00-0
:NaNO₂
:MFCD00011118