


Sodium nitrite ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7632-00-0
:
Formula:NaNO₂
MDL Number:MFCD00011118
:69 g/mol
Merck Index:14,8648
Boiling Pt:320 °C (1013 hPa)
Melting Pt:271 °C
Density:2.17
Storage Temperature:Ambient
EINECS:231-555-9
UN:1500
ADR:5.1,III
:GHS03
:GHS06
:GHS09
Related Information
Frequently Bought Together