


Sodium nitrite 97+% ACS
:7632-00-0
:
:NaNO₂
:MFCD00011118
:69 g/mol
:13,08721
:320 °C (1013 hPa)
:271 °C
:2,168 g/cm³ (20 °C)
:Ambient
:231-555-9
:1500
:5.1,III
:GHS06
:GHS09
:GHS03
- Appearance:White to light yellow Crystalline powder or crystals
- Titration with KMnO4:≥97.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤100 ppm
- Calcium (Ca):≤100 ppm
- Iron (Fe):≤10 ppm
- Potassium (K):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.01 %
Frequently Bought Together