Sodium perchlorate, anhydrous 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7601-89-0
:
Formula:ClNaO₄
MDL Number:MFCD00003480
:122.44 g/mol
Merck Index:14,8653
Melting Pt:482 °C (dec.)
Density:2.50
Storage Temperature:Ambient
EINECS:231-511-9
UN:1502
ADR:5.1,II
:GHS03
:GHS07
Related Information
Frequently Bought Together