Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
Potassium chlorate ≥99.0%, AnalaR NORMAPUR® analytical reagent
  26746.296
 :  
undefined
 
 
 
Potassium chlorate ≥99.0%, AnalaR NORMAPUR® analytical reagent
  26746.296
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    Min. 99.0 %
  • Acidity or alkalinity:
    Max. 0.002 meq/g
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Total N (Nitrogen):
    Max. 100 ppm
  • BrO3 (Bromate):
    Max. 150 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Ba (Barium):
    Max. 20 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Na (Sodium):
    Max. 100 ppm

 

 

 :
 :122.55 g/mol
 :13,07703
 :400 °C (1013 hPa)
 :356 °C
 :2.34 g/cm³ (20 °C)
 :Ambient
 :223-289-7
 :1485
 :5.1,II
 :GHS03
 :GHS07
 :GHS09
 :3811-04-9
 :KClO₃
 :MFCD00011361