


Potassium chlorate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3811-04-9
:
Formula:KClO₃
MDL Number:MFCD00011361
:122.55 g/mol
Merck Index:13,07703
Boiling Pt:400 °C (1013 hPa)
Melting Pt:356…368 °C
Density:2,34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:223-289-7
UN:1485
ADR:5.1,II
:GHS07
:GHS09
:GHS03
Specification Test Results
- Appearance:White Crystalline powder
- Assay:≥98.5 % (KClO3)
- Chloride (Cl):≤2000 ppm
- Bromate (BrO3):≤100 ppm
- Chromium (Cr):≤200 ppm
- Iron (Fe):≤20 ppm
- Insoluble matter:≤0.2 %
Specification Test Results
Frequently Bought Together