Magnetic stirrer heads, BOLA
Supplier: Bohlender
Certificates
About this item
Magnetic stirrer heads consist of perfect combination of drive shaft with ball bearings, rotor and lower bearing made of PTFE and a conductor made of borosilicate glass.
- No grease required
- Safe to run dry
- Long durability and compact size
- No leakage or memory effects due to non-porous, welded rotor
- Temperature range: −100 to 240 °C
- Chemical resistance
The rotor of this unit holds the stirrer shaft by means of three stud screws which are fixed in the counterbores of the stirrer shaft. The 6 mm square can be fixed into the stirrer coupling or into the agitator.
The outer Ø of space saving drive shaft is measured as 28 mm and 38 mm for NS 19/26 and NS 29/32 or NS 45/40 respectively.
Specifications
- Catalog No:
- BOHLC502-08
- BOHLC512-08
- Speed range:
- 1500 min⁻¹
- 1500 min⁻¹