Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
Boiling Pt:1413
Melting Pt:801
Density:2.16 g/cm³ (20 °C)
Molecular Weight:58.44 g/mol
Merck Index:14,08599
Storage Temperature:Ambient
EINECS:231-598-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.5 - 100.5 %
- Appearance of Solution S:Passes test Ph.Eur.
- Identification A:Passes test Ph.Eur.
- Identification B:Passes test Ph.Eur.
- NO2 (Nitrite):Passes test Ph.Eur.
- Solution in water:Passes test
- Acidity or alkalinity:Max. 0.0005 meq/g
- pH (25°C; 5 %):5.0 - 9.0
- Heavy metals (as Pb):Max. 2 ppm
- Hexacyanoferrates (as Fe(CN)6):Max. 1 ppm
- Insolubility in water:Max. 50 ppm
- Loss on drying (105°C):Max. 0.5 %
- Magnesium and alkaline earth metals:Max. 100 ppm
- Total N (Nitrogen):Max. 5 ppm
- Br (Bromide):Max. 50 ppm
- ClO3 + NO3 (as NO3):Max. 30 ppm
- I (Iodide):Max. 10 ppm
- PO4 (Phosphate):Max. 5 ppm
- SO4 (Sulphate):Max. 40 ppm
- As (Arsenic):Max. 0.4 ppm
- Ba (Barium):Max. 10 ppm
- Ca (Calcium):Max. 20 ppm
- Cu (Copper):Max. 2 ppm
- Fe (Iron):Max. 1 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 10 ppm
- Ni (Nickel):Max. 10 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC27810.262 spec_VWRC27810.262 spec_VWRC27810.295 spec_VWRC27810.295 spec_VWRC27810.364
Product specification
More
Frequently Bought Together