Potassium chloride, Optical Grade Optical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Potassiumchloride
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
Boiling Pt:1500 °C (subl.)
Melting Pt:773
Density:1.984
Molecular Weight:74.55 g/mol
Merck Index:14,7621
Storage Temperature:Ambient
EINECS:231-211-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together