Order Entry
Netherlands
ContactUsLinkComponent
Potassium hydrogen carbonate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Potassium hydrogen carbonate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 26733.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydrogen carbonate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 26733.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Substances reducing iodine (0,1 N):
    Passes test
  • pH (20°C; 5 %):
    8.2 - 8.8
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Substances precipitated by NH4OH:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • NH4 (Ammonium):
    Max. 10 ppm
  • NO3 (Nitrate):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Ca + Mg (as Ca):
    Max. 100 ppm
  • Fe (Iron):
    Max. 2 ppm
  • Na (Sodium):
    Max. 0.025 %

Specification Test Results

About this item

 :
 :100.12 g/mol
Merck Index:13,07691
Melting Pt:292 °C
Density:2.17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-059-0
CAS Number:298-14-6
Formula:KHCO₃
MDL Number:MFCD00011402