Potassium hydrogen carbonate FCC, E501, SAFC®
Catalog # 1.72574.9025
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:100.12 g/mol
Merck Index:13,07691
Melting Pt:292 °C
Density:2.17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-059-0
CAS Number:298-14-6
Formula:KHCO₃
MDL Number:MFCD00011402