Order Entry
Netherlands
ContactUsLinkComponent
Potassium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent sodium-free
Irritant
Potassium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent sodium-free
Catalog # 26727.267
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Potassium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent sodium-free
Catalog # 26727.267
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    Min. 99.0 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (300 °C):
    Max. 1 %
  • Substances precipitated by NH4OH:
    Max. 100 ppm
  • Total N (Nitrogen):
    Max. 10 ppm
  • Total S (as SO4):
    Max. 30 ppm
  • Cl (Chloride):
    Max. 30 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • SiO4 (as SiO2):
    Max. 50 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Mg (Magnesium):
    Max. 10 ppm
  • Na (Sodium):
    Max. 0.02 %
  • Pb (Lead):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :138.21 g/mol
Merck Index:13,07702
Melting Pt:891 °C
Density:2.42 g/cm³ (20 °C)
EINECS:209-529-3
 :GHS07
Storage Temperature:Ambient
CAS Number:584-08-7
Formula:K₂CO₃
MDL Number:MFCD00011382