Supelco® Organic Reference Standard, Cyclohexene
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P240 - P273 - P301 + P310 - P331
Specifications
- CAS No.:110-83-8
- Storage conditions:2...30 °C
- Beschrijving van de gevarenklasse:H225 - H302 - H304 - H411
- Shipping conditions:STD
- MDL No.:MFCD00001539
- M-Clarity quality level:MQ100
- Synonym(s):Tetrahydrobenzene
- Smeltpuntapparaat:–104 °C (lit.)
- Moleculair gewicht:82,14
- Dichtheid:0,811 g/ml at 25 °C (lit.)
- Vlampunt:–12 °C - closed cup
- Kookpunt:83 °C (lit.)
Specifications
Frequently Bought Together