
3-Picolyl bromide hydrobromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4916-55-6
:
Formula:C₆H₇Br₂N
MDL Number:MFCD01321314
:252.94 g/mol
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together