Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, 2,4,5-Trimethylaniline 100 µg/ml in acetonitrile, Supelco®
Organic reference standard, 2,4,5-Trimethylaniline 100 µg/ml in acetonitrile, Supelco®
Catalog # 31039-2ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2,4,5-Trimethylaniline 100 µg/ml in acetonitrile, Supelco®
Catalog # 31039-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • CAS No.:
    137-17-7
  • Description:
    2,4,5-Trimethylaniline 100 µg/ml in acetonitrile
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₉H₁₃N
  • Description:
    2,4,5-Trimethylaniline

Specifications

About this item