Pesticide reference standard, 2,6-Dichlorobenzamide 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA12355000AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:2008-58-4
- Description:2,6-Dichlorobenzamide 100 µg/ml in acetonitrile
- Formula:C₇H₅Cl₂NO
- Description:2,6-Dichlorobenzamide