
Methyl-3-(trifluoromethyl)benzoate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2557-13-3
:
Formula:C₉H₇F₃O₂
MDL Number:MFCD00043543
:204.15 g/mol
Boiling Pt:80…82 °C (15 mmHg)
Density:1.295
Flash Pt:84 °C (183 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together