
Ytterbium(III) trifluoromethanesulphonate hydrate 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:252976-51-5
:54761-04-5
Formula:C₃F₉O₉S₃Yb·xH₂O
MDL Number:MFCD03703499
:620.25 g/mol (anhy)
Melting Pt:118…122 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White Crystalline powder
- Total conc. of imp.as REO:≤0.1 %
- Ytterbium (Yb):20.0 to 26.0 %
Specification Test Results
Frequently Bought Together