
DL-Tropic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:552-63-6
Formula:C₉H₁₀O₃
MDL Number:MFCD00004255
:166.18 g/mol
Melting Pt:117…120 °C
EINECS:208-465-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:115 to 120 °C
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together