1,3-Cyclohexanedione
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: Cyclohexane-1,3-dione
CAS Number:504-02-9
:
Formula:C₆H₈O₂
MDL Number:MFCD00001585
Boiling Pt:235
Melting Pt:105
Density:1.25 g/cm³ (20 °C)
Molecular Weight:112.13 g/mol
Merck Index:13,03200
Storage Temperature:Refrigerator
EINECS:207-980-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together