Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Clioquinol
Clioquinol
Clioquinol
PROOBP084
:
LGC Standards PROMOCHEM
:
Clioquinol
PROOBP084
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
200 mg
Seq:
2530
:
:
12,05052
:
365 °C (1013 hPa)
:
183…184 °C
:
Ambient
:
6.1,III
:
305.5
:
204-984-4
:
2811
:
GHS06
:
130-26-7
:
C₉H₅ClINO
:
MFCD00006787