Order Entry
Netherlands
Orders LinkContactUsLinkComponent
m-Xylene ≥99%

m-Xylene ≥99%

Flammable
Irritant
m-Xylene ≥99%
Certificates
m-Xylene ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 108-38-3 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 25°(77°F) Refractive Index: 1.4970 Odour: Mild Melting Point: -47° Boiling Point: 138-139° Beilstein Reference: 605441 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00008536

Synonyms:  1,3-Dimethylbenzene
  • CAS Number:108-38-3
  • Formula:C₈H₁₀
  • MDL Number:MFCD00008536
  • Molecular Weight:106.17 g/mol
  • Merck Index:14,10081
  • Boiling Pt:138…139 °C
  • Melting Pt:–47 °C
  • Density:0.866
  • Flash Pt:25 °C (77 °F)
  • Storage Temperature:Ambient
  • EINECS:203-576-3
  • UN:1307
  • ADR:3,II
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.