


m-Xylene 99+%, extra pure
:108-38-3
:
:C₈H₁₀
:MFCD00008536
:106.17 g/mol
:12,10214
:138 °C (1013 hPa)
:–48 °C
:0.864 g/cm³
:25 °C
:Ambient
:203-576-3
:1307
:3,III
:GHS02
:GHS07
:GHS08
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4960 to 1.4980 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥99.0 %
- Residue after evaporation:≤0.005 %
Frequently Bought Together