


2,6-Xylenol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:576-26-1
:
Formula:C₈H₁₀O
MDL Number:MFCD00002240
:122.17 g/mol
Merck Index:12,10215
Boiling Pt:203 °C (1013 hPa)
Melting Pt:41…42 °C
Density:1.13 g/cm³ (20 °C)
EINECS:209-400-1
UN:2261
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:73 °C
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:8 g/l (20 °C)
- Melting Point:45 - 48 °C
- Molar Mass:122.16 g/mol
- Boiling Point:203 °C
- Vapor Pressure:0.2 hPa (20 °C)
- Flash Point:73 °C
- Explosion Limit:1.4 %(V)
- Density:1.03 g/cm3 (20 °C)
- pH:6 - 7 (8 g/l, H2O, 20 °C)
- Ignition Point:555 °C
Specification Test Results
Frequently Bought Together